Learn More
Rbin-1, MedChemExpress
Product Code. 16480699
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480699 1 mg 1mg
16475907 5 mg 5mg
16469726 50 mg 50mg
16453178 10 mg 10mg
16432656 25 mg 25mg
16414466 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16480699 Supplier Medchem Express Supplier No. HY100816
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Rbin-1 is a potent, reversible, and specific chemical inhibitor of eukaryotic ribosome biogenesis. Rbin-1 inhibits the ATPase with GI50 of 136 nM. Rbin-1 is a potent and selective chemical inhibitor of Midasin (Mdn1).
Rbin-1 is a potent, reversible, and specific chemical inhibitor of eukaryotic ribosome biogenesis. Rbin-1 inhibits the ATPase with GI50 of 136 nM. Rbin-1 is a potent and selective chemical inhibitor of Midasin (Mdn1).
Specifications
Chemical Name or Material Rbin-1
Physical Form Solid
CAS 328023-11-6
Color Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C13H12N4S
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym Ribozinoindole-1
Solubility Information DMSO : ≥ 31 mg/mL (120.94 mM)
SMILES C=C(C)CSC1=NN=C2C(NC3=C2C=CC=C3)=N1
Molecular Weight (g/mol) 256.33
Formula Weight 256.33
Percent Purity 99.33%
Grade Research