Learn More
Rauwolscine hydrochloride, 99%
Product Code. 15475459
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15475459 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15475459 Supplier Thermo Scientific Alfa Aesar Supplier No. J63198.MC
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 6211-32-1 | C21H27ClN2O3 | 390.908 g/mol
Standard alpha-2 adrenergic antagonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 6211-32-1
Color White
Molecular Formula C21H27ClN2O3
MDL Number MFCD06668052
Quantity 100 mg
UN Number UN1544
Sensitivity Light sensitive
Synonym 17?-hydroxy-20?-yohimban-16?-carboxylic acid,methyl ester hydrochloride
InChI Key PIPZGJSEDRMUAW-XQDOVXNESA-N
SMILES COC(=O)C1C(CCC2C1CC3C4=C(CCN3C2)C5=CC=CC=C5N4)O.Cl
IUPAC Name methyl (1S,15S,18S,19S)-18-hydroxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate;hydrochloride
Molecular Weight (g/mol) 390.908
PubChem CID 90479742
Formula Weight 390.91
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Rauwolscine hydrochloride