missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Fas/TNFRSF6/CD95 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF2159
This item is not returnable.
View return policy
Description
Fas/TNFRSF6/CD95 Polyclonal specifically detects Fas/TNFRSF6/CD95 in Rat samples. It is validated for Western Blot.
Specifications
| Fas/TNFRSF6/CD95 | |
| Polyclonal | |
| Biotin | |
| NP_631933 | |
| FAS | |
| Mouse myeloma cell line NS0-derived recombinant rat Fas/TNFRSF6/CD95 Gln22-Lys170 Accession # NP_631933 | |
| 50 μg | |
| Primary | |
| Detects rat Fas/TNFRSF6/CD95 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant human Fas, recombinant mouse Fas, and recombinant feline Fas (under non-reducing conditions) is observed. | |
| Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Apo-1, Apo-1 antigen, apoptosis antigen 1, Apoptosis-mediating surface antigen FAS, APT1, APT1FASTM, CD95, CD95 antigen, CD95ALPS1A, Fas (TNF receptor superfamily, member 6), Fas AMA, Fas antigen, FAS1, FASLG receptor, TNFRSF6, TNFRSF6member 6, tumor necrosis factor receptor superfamily member 6 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 355 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction