missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL2, Rat, Clone: I2-C7, Abnova™
Rat monoclonal antibody raised against native Il2.
Brand: Abnova MAB1324
This item is not returnable.
View return policy
Specifications
| IL2 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Il2 | |
| Il2 | |
| Native purified Il2 from mouse Con A activated T cell culture supenatant. | |
| RUO | |
| 16183 | |
| Mouse | |
| Lyophilized |
| ELISA | |
| I2-C7 | |
| Rat monoclonal antibody raised against native Il2. | |
| Lyophilized from PBS | |
| Il-2 | |
| Rat | |
| 200 μg | |
| Primary | |
| React with mouse Il2. Does not cross react with rat or human IL2. | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction