missing translation for 'onlineSavingsMsg'
Learn More

Rat Agrin Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18041902
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18041902 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18041902 Supplier R&D Systems Supplier No. AF550

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

Agrin Polyclonal specifically detects Agrin in Rat samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Agrin
Applications Western Blot, Immunohistochemistry, Neutralization, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.001-0.004 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Gene Accession No. P25304
Gene Alias agrin, agrin proteoglycan, AGRN
Gene Symbols AGRN
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant rat Agrin Ala1153-Pro1959 (Pro1788-Ser1798 del) Accession # P25304
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375790
Test Specificity Detects rat Agrin in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant rat (rr) GDNF, rrIL-1 alpha , and rrL-Selectin is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.