missing translation for 'onlineSavingsMsg'
Learn More

RAPH1 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16824315 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16824315 20 μL 20µL
16814315 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16824315 Supplier Proteintech Supplier No. 257501AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes the L protein of the mitochondrial glycine cleavage system. The L protein, also named dihydrolipoamide dehydrogenase, is also a component of the pyruvate dehydrogenase complex, the alpha-ketoglutarate dehydrogenase complex, and the branched-chain alpha-keto acide dehydrogenase complex. Mutations in this gene have been identified in patients with E3-deficient maple syrup urine disease and lipoamide dehydrogenase deficiency.

Specifications

Antigen RAPH1
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RAPH1
Gene Accession No. Q70E73
Gene Alias ALS2CR18, ALS2CR9, KIAA1681, Lamellipodin, LPD, PREL 2, PREL2, Proline rich EVH1 ligand 2, Protein RMO1, RalGDS/AF 6, RAPH1, RMO1
Gene Symbols RAPH1
Host Species Rabbit
Immunogen RAPH1 Fusion Protein Ag22705
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65059
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.