missing translation for 'onlineSavingsMsg'
Learn More

Rapastinel Trifluoroacetate, MedChemExpress

Product Code. 16417168
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16417168

Brand: Medchem Express HY16728B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rapastinel Trifluoroacetate (GLYX-13 Trifluoroacetate) is an NMDA receptor modulator with glycine-site partial agonist properties. Rapastinel Trifluoroacetate has the potential for major depressive disorder treatment.

Rapastinel Trifluoroacetate (GLYX-13 Trifluoroacetate) is an NMDA receptor modulator with glycine-site partial agonist properties. Rapastinel Trifluoroacetate has the potential for major depressive disorder treatment.

Specifications

Chemical Name or Material Rapastinel Trifluoroacetate
CAS 1435786-04-1
Color White
Purity Grade Notes Research
Molecular Formula C20H32F3N5O8
Quantity 10 mg
Synonym GLYX-13 Trifluoroacetate
Solubility Information DMSO : 125 mg/mL (236.97 mM; Need ultrasonic)
SMILES O=C([C@H]1N(C([C@H]2N(C([C@@H](N)[C@H](O)C)=O)CCC2)=O)CCC1)N[C@@H]([C@H](O)C)C(N)=O.O=C(O)C(F)(F)F
Molecular Weight (g/mol) 527.49
Formula Weight 527.49
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.