missing translation for 'onlineSavingsMsg'
Learn More

RAMP3 Antibody (Medella patent anti-RAMP-3), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 30116180 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30116180 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30116180 Supplier Novus Biologicals Supplier No. NBP327994MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

RAMP3 Monoclonal antibody specifically detects RAMP3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAMP3
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone Medella patent anti-RAMP-3
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias calcitonin receptor-like receptor activity modifying protein 3, Calcitonin-receptor-like receptor activity-modifying protein 3, CRLR activity-modifying protein 3, receptor (calcitonin) activity modifying protein 3, receptor (G protein-coupled) activity modifying protein 3, receptor activity modifying protein 3, receptor activity-modifying protein 3, receptor-activity-modifying protein 3
Host Species Human
Immunogen RAMP3
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10268
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.