Learn More
Ohaus™ Ramp 800 VFS
Product Code. 15460961 Shop All Ohaus Products
Change view
Click to view available options
Weight: 36.700 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15460961 Weight: 36.700 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15460961 Supplier Ohaus™ Supplier No. 22015473
Please to purchase this item. Need a web account? Register with us today!
OHAUS™ Ramp 800 VFS is designed to be used with a variety of floor scales and combi floor scales.
  • Accessory
  • For use with Floor Scale VFS-CS300 and VFS-CS600
  • For use with Combi Floor Scale VFS-CS30051XW, VFS-CS30051XWM, VFS-CS60051XW, and VFS-CS60051XWM
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Floor Scale and Combi Floor Scale
Color Gray
Languages Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX)
Weight (Metric) 36.700 kg