missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Raloxifene (Hydrochloride)

Product Code. 17180132
Encompass_Preferred
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 17180132

Brand: STEMCELL Technologies 72854

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective estrogen receptor modulator

  • Raloxifene is a selective estrogen receptor modulator (SERM) that exhibits agonistic (estrogenic) activity in bone cells without stimulating ER activity in breast or uterine tissues (Black et al.)
  • Enhances osteoblast differentiation in mouse bone marrow and human osteoblast cultures (Qu et al.; Viereck et al.)
  • Inhibits osteoclast differentiation in primary human bone marrow mononuclear cell cultures (Ramalho et al.)
  • Reduced endodermal differentiation (HNF-4α expression) in embryoid bodies derived from human embryonic stem (ES) cells (Kim et al.)

Specifications

CAS 82640-04-8
Synonym Evista™; Keoxifene; LY156758
Quantity 500 mg
Molecular Formula C28H27NO4S · HCl
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.