Learn More
RAF265, MedChemExpress
Product Code. 16490117
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16490117
Brand: Medchem Express HY10248
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress RAF265 is a potent RAF/VEGFR2 inhibitor.
RAF265 is a potent RAF/VEGFR2 inhibitor.
Specifications
Chemical Name or Material RAF265
CAS 927880-90-8
Color White
Purity Grade Notes Research
Molecular Formula C24H16F6N6O
Quantity 50 mg
Synonym CHIR-265
Solubility Information DMSO : ≥ 26 mg/mL (50.15 mM) ∣Ethanol : 10 mg/mL (19.29 mM; Need ultrasonic)
SMILES FC(F)(C1=CN=C(N1)C2=NC=CC(OC3=CC=C4N(C(NC5=CC=C(C(F)(F)F)C=C5)=NC4=C3)C)=C2)F
Molecular Weight (g/mol) 518.41
Formula Weight 518.41
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease