Learn More
Invitrogen™ RAE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA593166
Description
Immunogen sequence: MSLFGTTSGF GTSGTSMFGS ATTDNHNPMK DIEVTSSPDD SIGCLSFSPP TLPGNFLIAG SWANDVRCWE VQDSGQTIPK AQQMHTGPVL DVCWSDDGSK VFTASCDKTA KMWDLSSNQA IQIAQHDAPV KTIHWIKAPN YSCVMTGSWD KTLKFWDTRS SNPMMVLQLP ERCYCADVIY PMAVVATAER GLIVYQLENQ PSEFRRIESP LKHQHRCVAI FKDKQNKPTG FALGSIEGRV AIHYINPPNP AKDNFTFKCH RSNGTNTSAP QDIYAVNGIA FHPVHGTLAT VGSDGRFSFW DKDARTKLKT SEQLDQPISA CCFNHNGNIF AYASSYDWSK GHEFYNPQKK NYIFLRNAAE ELKPRNKK; Positive Samples: Mouse brain, Mouse spleen, Mouse thymus, Rat brain; Cellular Location: Cytoplasm, Nucleus.
Mutations in the Schizosaccharomyces pombe Rae1 and Saccharomyces cerevisiae Gle2 genes have been shown to result in accumulation of poly-containing mRNA in the nucleus, suggesting that the encoded proteins are involved in RNA export. The protein encoded by this gene is a homolog of yeast Rae1. It contains four WD40 motifs, and has been shown to localize to distinct foci in the nucleoplasm, to the nuclear rim, and to meshwork-like structures throughout the cytoplasm. This gene is thought to be involved in nucleocytoplasmic transport, and in directly or indirectly attaching cytoplasmic mRNPs to the cytoskeleton. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| RAE1 | |
| Polyclonal | |
| Unconjugated | |
| RAE1 | |
| 3230401I12Rik; 41; D2Ertd342e; dJ481F12.3; dJ800J21.1; FLJ30608; HGNC:9828; homolog of yeast Rae1 (Bharathi) mRNA-associated protein of 41 kDa (Kraemer); MGC117333; MGC126076; MGC126077; MIG14; migration-inducing gene 14; MNRP; Mnrp41; mRNA export factor; mRNA export factor-like protein; mRNA export protein; mRNA-associated protein MRNP 41; mRNA-binding protein, 41-kD; MRNP41; OTTHUMP00000166110; rae1; RAE1 (RNA export 1, S.pombe) homolog; rae1 protein; rae1 protein homolog; RAE1 RNA export 1 homolog; RAE1 RNA export 1 homolog (S. pombe); ribonucleic acid export 1; RP4-800J21.2; zgc:56449; zgc:77723 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 362281, 66679, 8480 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.67 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P78406, Q3SWS8, Q8C570 | |
| RAE1 | |
| Recombinant Protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.