missing translation for 'onlineSavingsMsg'
Learn More

Radezolid, MedChemExpress

Product Code. p-200100937
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16438777

Brand: Medchem Express HY14800

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Radezolid (RX-1741) is a oxazolidinone antibiotic. Radezolid is active against Staphylococcus, Chlamydia, and Legionella species, and remains active against Linezolid-resistant strains.

Radezolid (RX-1741) is a oxazolidinone antibiotic. Radezolid is active against Staphylococcus, Chlamydia, and Legionella species, and remains active against Linezolid-resistant strains.

Specifications

Chemical Name or Material Radezolid
Physical Form Solid
CAS 869884-78-6
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H23FN6O3
For Use With (Application) COVID-19-immunoregulation
Synonym RX-1741
Solubility Information DMSO : ≥ 25 mg/mL (57.02 mM)
SMILES CC(NC[C@H]1CN(C2=CC=C(C3=CC=C(CNCC4=CN=NN4)C=C3)C(F)=C2)C(O1)=O)=O
Molecular Weight (g/mol) 438.45
Formula Weight 438.45
Percent Purity 98.02%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.