missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD54L Antibody (Rad54 4E3/1), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-13364H
This item is not returnable.
View return policy
Description
RAD54L Monoclonal specifically detects RAD54L in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| RAD54L | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| RAD54L | |
| Full length human RAD54L protein | |
| 0.1 mL | |
| DNA Repair, Homologous Recombination | |
| 8438 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| Rad54 4E3/1 | |
| Western Blot, Immunohistochemistry | |
| EC 3.6.1, hHR54EC 3.6.4.-, HR54, hRAD54HRAD54, RAD54 (S.cerevisiae)-like, RAD54 homolog, RAD54ADNA repair and recombination protein RAD54-like, RAD54-like (S. cerevisiae) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction