Learn More
Edmund Bühler GmbH™ Combifix VKS C Rack System
Includes 2 clamping strips h, 2 cramp strips and 1 spring strip. For fastening separating funnels.
Brand: Edmund Bühler GmbH™ 0051 489
Additional Details : Weight : 1.00000kg
Description
- Basic rack system includes:
- 2 clamping strips h
- 2 cramp strips
- 1 spring strip
- For use with fastening separating funnels.
- Standard version of cramp strip: 7 clamps NS 29 (D32)
- To be mounted on the shaking plate.
Specifications
Combifix VKS C Rack System | |
Combifix VKS | |
2 clamping strips H, 2 cramp strips, 1 spring strip |