Learn More
Tenak™ Microtiterplates Comfort Rack For Upright Freezers
Shop All TENAK Products
Click to view available options
Holds:
Plates (138)
Plates (48)
Plates (56)
Plates (70)
Plates (90)
Plates (91)
Plates (96)
Height (Metric) Shelf:
16 mm
25 mm
45 mm
Unit Size:
Each
Description
- Variety of shelf heights available
- Racks can either be used as one plate per shelf or the plates can be stacked
- Includes an alert preventing the shelves from sliding out
Specifications
Specifications
| Product Type | Microtiterplate Rack |
| Array | 13 x 7 |
| Holds | Plates (91) |
| For Use With (Equipment) | Upright Freezers |
| Height (Metric) Shelf | 16 mm |