Learn More
Tenak™ Classic Rack for Chest Freezers, with Locking Rod, 32 mm Box height
Classic Rack is suitable for use with all chest freezer models.
Brand: Tenak™ TE21358
Additional Details : Weight : 2.00000kg
Description
- Comes with a locking rod to secure the cryoboxes
- Measure the inside height of you freezer when choosing the right rack
- Constructed of stainless steel
Specifications
Stainless Steel | |
Boxes (16) | |
140 x 140 x 572 mm |
Chest Frseezers | |
Locking rod | |
32 mm |