Learn More
rac Viloxazine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Viloxazine Hydrochloride |
| CAS | 35604-67-2 |
| Molecular Formula | C13 H19 N O3 . Cl H |
| Quantity | 250 mg |
| Synonym | Morpholine','2-[(2-ethoxyphenoxy)methyl]-','hydrochloride (1:1), Morpholine','2-[(2-ethoxyphenoxy)methyl]-','hydrochloride (9CI), 2-[(2-Ethoxyphenoxy)methyl]morpholinium chloride, Catatrol, ICI 58834, Vicilan, Viloxazin hydrochloride, Viloxazine hydrochloride, Vivalan, Vivarint, Vivilan |
| InChI Formula | InChI=1S/C13H19NO3.ClH/c1-2-15-12-5-3-4-6-13(12)17-10-11-9-14-7-8-16-11;/h3-6,11,14H,2,7-10H2,1H3;1H |
| SMILES | Cl.CCOc1ccccc1OCC2CNCCO2 |
| IUPAC Name | 2-[(2-ethoxyphenoxy)methyl]morpholine;hydrochloride |
| Molecular Weight (g/mol) | 273.76 |
| Formula Weight | 273.1132 |