missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rac Styrene Glycol, TRC
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | Styrene Glycol |
| CAS | 93-56-1 |
| Molecular Formula | C8 H10 O2 |
| Quantity | 1 g |
| Synonym | 1,2-Ethanediol','1-phenyl-, 1,2-Ethanediol','phenyl- (6CI,7CI), 1-Phenyl-1,2-ethanediol, (1,2-Dihydroxyethyl)benzene, (RS)-1-Phenyl-1,2-ethanediol, (±)-1-Phenyl-1,2-ethanediol, (±)-Phenyl glycol, (±)-Phenyl-1,2-ethanediol, (±)-Styrene glycol, 1,2-Dihydroxy-1-phenylethane, 1,2-Dihydroxy-2-phenylethane, 1-Phenyl-1,2-glycol, 1-Phenylethylene glycol, NSC 406601, Phenyl glycol, Phenylethylene glycol, Styrene glycol, Styrolyl alcohol, cis-1-Phenyl-1,2-ethanediol, α,β-Dihydroxyethylbenzene, β-Hydroxy-β-phenylethanol, 1-Phenylethane-1,2-diol |
| InChI Formula | InChI=1S/C8H10O2/c9-6-8(10)7-4-2-1-3-5-7/h1-5,8-10H,6H2 |
| SMILES | OCC(O)c1ccccc1 |
| IUPAC Name | 1-phenylethane-1,2-diol |
| Molecular Weight (g/mol) | 138.16 |
| Formula Weight | 138.0681 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction