missing translation for 'onlineSavingsMsg'
Learn More

(Rac)-Rotigotine hydrochloride, MedChemExpress

Product Code. 16412587
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16412587

Brand: Medchem Express HY15394

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (Rac)-Rotigotine hydrochloride is a racemate of Rotigotine. Rotigotine is a full agonist of dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor, with Kis of 0.71 nM, 4-15 nM, and 83 nM for the dopamine D3 receptor and D2, D5, D4 receptors, and dopamine D1 receptor.

(Rac)-Rotigotine hydrochloride is a racemate of Rotigotine. Rotigotine is a full agonist of dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor, with Kis of 0.71 nM, 4-15 nM, and 83 nM for the dopamine D3 receptor and D2, D5, D4 receptors, and dopamine D1 receptor.

Specifications

Chemical Name or Material (Rac)-Rotigotine hydrochloride
CAS 102120-99-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H26ClNOS
Quantity 50 mg
Solubility Information DMSO : ≥ 50 mg/mL (142.07 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES OC1=C2CCC(N(CCC)CCC3=CC=CS3)CC2=CC=C1.[H]Cl
Molecular Weight (g/mol) 351.93
Formula Weight 351.93
Percent Purity 98.66%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.