missing translation for 'onlineSavingsMsg'
Learn More

rac Desfluoro Citalopram Oxalate, TRC

Product Code. 30009889
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009889 25 mg 25mg
30136293 5 mg 5mg
30003545 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009889 Supplier Toronto Research Chemicals Supplier No. TRCD28996525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1093072-86-6
Formula Weight 396.1685
InChI Formula InChI=1S/C20H22N2O.C2H2O4/c1-22(2)12-6-11-20(18-7-4-3-5-8-18)19-10-9-16(14-21)13-17(19)15-23-20;3-1(4)2(5)6/h3-5,7-10,13H,6,11-12,15H2,1-2H3;(H,3,4)(H,5,6)
IUPAC Name 1-[3-(dimethylamino)propyl]-1-phenyl-3H-2-benzofuran-5-carbonitrile;oxalic acid
Molecular Formula C20 H22 N2 O . C2 H2 O4
Molecular Weight (g/mol) 396.44
Percent Purity >95%
SMILES CN(C)CCCC1(OCc2cc(ccc12)C#N)c3ccccc3.OC(=O)C(=O)O
Synonym 1-(3-Dimethylaminopropyl)-1-(phenyl)-5-phthalan-carbonitrile Oxalate, 1-[3-(Dimethylamino)propyl]-1,3-dihydro-1-phenyl-5-isobenzofurancarbonitrile Ethanedioate (1:1)
Chemical Name or Material Desfluoro Citalopram Oxalate
Grade HPLC
Show More Show Less

Specifications

Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.