Learn More
rac-Clopidogrel Hydrochloride, TRC
Product Code. 17944615
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17944615 10 mg 10mg
17950445 5 mg 5mg
17993395 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17944615 Supplier Toronto Research Chemicals Supplier No. TRCC58726010MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 144750-52-7
Molecular Formula C16 H16 Cl N O2 S . Cl H
Molecular Weight (g/mol) 358.28
Synonym Thieno[2,3-c]pyridine-6(5H)-acetic acid, α-(2-chlorophenyl)-4,7-dihydro-, methyl ester, hydrochloride (1:1), Thieno[2,3-c]pyridine-6(5H)-acetic acid, α-(2-chlorophenyl)-4,7-dihydro-, methyl ester, hydrochloride (9CI), Thieno[2,3-c]pyridine-6(5H)-acetic acid, α-(2-chlorophenyl)-4,7-dihydro-, methyl ester, hydrochloride, (±)-, Methyl (2RS)-(2-Chlorophenyl)[4,7-dihydrothieno[2,3-c]pyridin-6(5H)-yl]acetate hydrochloride, Methyl(±)-(o-chlorophenyl)-4,5-dihydrothieno[2,3-c]pyridine-6(7H)-acetate hydrochloride
IUPAC Name methyl 2-(2-chlorophenyl)-2-(5,7-dihydro-4H-thieno[2,3-c]pyridin-6-yl)acetate;hydrochloride
SMILES Cl.COC(=O)C(N1CCc2ccsc2C1)c3ccccc3Cl
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C16H16ClNO2S.ClH/c1-20-16(19)15(12-4-2-3-5-13(12)17)18-8-6-11-7-9-21-14(11)10-18;/h2-5,7,9,15H,6,8,10H2,1H3;1H
Formula Weight 357.0357
Percent Purity >95%
Chemical Name or Material Methyl (2-Chlorophenyl)[4,7-dihydrothieno[2,3-c]pyridin-6(5H)-yl]acetate Hydrochloride
Grade HPLC