Learn More
rac-5-Bromo Naproxen, TRC
Specifications
Specifications
| Chemical Name or Material | (±)-5-Bromonaproxen |
| CAS | 27655-95-4 |
| Molecular Formula | C14 H13 Br O3 |
| Quantity | 250 mg |
| Synonym | 2-Naphthaleneacetic acid, 5-bromo-6-methoxy-α-methyl-, 2-Naphthaleneacetic acid, 5-bromo-6-methoxy-α-methyl-, (±)- (8CI), 5-Bromo-6-methoxy-α-methyl-2-naphthaleneacetic acid, (±)-5-Bromo-6-methoxy-α-methyl-2-naphthaleneacetic acid, (±)-5-Bromonaproxen |
| InChI Formula | InChI=1S/C14H13BrO3/c1-8(14(16)17)9-3-5-11-10(7-9)4-6-12(18-2)13(11)15/h3-8H,1-2H3,(H,16,17) |
| SMILES | COc1ccc2cc(ccc2c1Br)C(C)C(=O)O |
| IUPAC Name | 2-(5-bromo-6-methoxynaphthalen-2-yl)propanoic acid |
| Molecular Weight (g/mol) | 309.16 |
| Formula Weight | 308.0048 |