Learn More
rac 4-Hydroxy Ephedrine Hydrochloride, TRC
Product Code. 17993575
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17993575 10 mg 10mg
17963285 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17993575 Supplier Toronto Research Chemicals Supplier No. TRCH82530510MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 942-51-8
Molecular Formula C10 H15 N O2 . Cl H
Molecular Weight (g/mol) 217.69
Synonym Oxilofrine hydrochloride, Benzenemethanol, 4-hydroxy-α-[1-(methylamino)ethyl]-, hydrochloride, (R*,S*)- (9CI), 4-Hydroxyephedrinehydrochloride, Suprifen hydrochloride, p-Hydroxyephedrine hydrochloride
IUPAC Name 4-[(1R,2S)-1-hydroxy-2-(methylamino)propyl]phenol;hydrochloride
SMILES Cl.CN[C@@H](C)[C@H](O)c1ccc(O)cc1
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C10H15NO2.ClH/c1-7(11-2)10(13)8-3-5-9(12)6-4-8;/h3-7,10-13H,1-2H3;1H/t7-,10-;/m0./s1
Formula Weight 217.087
Percent Purity >95%
Chemical Name or Material Oxilofrine Hydrochloride
Grade HPLC