Learn More
Rac 1,2-Dioleoyl-3-chloropropanediol-d5, TRC
Product Code. 30387479
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30387479 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30387479 Supplier Toronto Research Chemicals Supplier No. TRCD4822022.5MG
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Chloro-1,2-propanediol Dioleate D5 (3-Chloro-1,2-propandiol D5)
CAS 1246833-00-0
Purity Grade Notes HPLC
Molecular Formula C39 2H5 H66 Cl O4
Quantity 2.5 mg
Synonym 3-Chloro-1,2-propanediol Dioleate D5 (3-Chloro-1,2-propandiol D5)
InChI Formula InChI=1S/C39H71ClO4/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-36-37(35-40)44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20,37H,3-16,21-36H2,1-2H3/b19-17-,20-18-/i35D2,36D2,37D
SMILES [2H]C([2H])(Cl)C([2H])(OC(=O)CCCCCCC\C=C/CCCCCCCC)C([2H])([2H])OC(=O)CCCCCCC\C=C/CCCCCCCC
IUPAC Name [3-chloro-1,1,2,3,3-pentadeuterio-2-[(Z)-octadec-9-enoyl]oxypropyl] (Z)-octadec-9-enoate
Molecular Weight (g/mol) 644.46
Formula Weight 643.5355
Percent Purity >95