Learn More
RABGGTB Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | RABGGTB |
|---|---|
| Concentration | 267 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16879113
|
Proteintech
11801-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16869113
|
Proteintech
11801-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RABGGTB antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RABGGTB. This antibody recognizes human, mouse, rat antigen. The RABGGTB antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| RABGGTB | |
| Western Blot, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG |
| 267 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 5876 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.