Learn More
RNF44, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RNF44.
Brand: Abnova PAB23378.100uL
Description
The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. [provided by RefSeq
Sequence: MRPWALAVTRWPPSAPVGQRRFSAGPGSTPGQLWGSPGLEGPLASPPARDERLPSQQPPSRPPHLPVEERRASAPAGGSPRMLHPATQSpecifications
| RNF44 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RNF44. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| KIAA1100 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 22838 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| RNF44 | |
| RNF44 | |
| Recombinant protein corresponding to amino acids of human RNF44. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only