Learn More
PRKAA1 Rabbit anti-Bovine, Human, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PRKAA1.
Brand: Abnova PAB14475.100ug
Description
The protein encoded by this gene belongs to the ser/thr protein kinase family. It is the catalytic subunit of the 5'-prime-AMP-activated protein kinase (AMPK). AMPK is a cellular energy sensor conserved in all eukaryotic cells. The kinase activity of AMPK is activated by the stimuli that increase the cellular AMP/ATP ratio. AMPK regulates the activities of a number of key metabolic enzymes through phosphorylation. It protects cells from stresses that cause ATP depletion by switching off ATP-consuming biosynthetic pathways. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]
Specifications
| PRKAA1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PRKAA1. | |
| In Tris-citrate/phosphate, pH 7-8 (0.09% sodium azide) | |
| AMPK/AMPKa1/MGC33776/MGC57364 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Bovine, Human, Rat | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (0.2-2 ug/mL) Immunoprecipitation (2-8 ug/mg) The optimal working dilution should be determined by the end user. | |
| PRKAA1 | |
| PRKAA1 | |
| A synthetic peptide corresponding to the portion encoded within exon 8 of human PRKAA1. | |
| RUO | |
| 5562 | |
| Store at 4°C. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only