missing translation for 'onlineSavingsMsg'
Learn More

lacI, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against full-length recombinant lacI.

Brand:  Abnova PAB10255.100ug

Product Code. 16089136

  • 870.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

lacI
Polyclonal
Rabbit polyclonal antibody raised against full length recombinant lacI.
In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
ECK0342/JW0336
Rabbit
100 μg
Primary
This antibody reacts with recombinant Lac I protein. A BLAST analysis was used to suggest cross-reactivity with Lac I from DNA-binding transcriptional repressor in Escherichia coli (strain DH10B) and transcriptional repressor of the lac operon in Shigella sonnei (strain Ss046) based on a 100% homology with the immunizing sequence.
Liquid
ELISA, Western Blot
Unconjugated
ELISA (1:90000-1:100000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
lacI
lacI
Recombinant protein corresponding to full length Escherichia coli (strain DH10B) lacI.
RUO
945007
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only