missing translation for 'onlineSavingsMsg'
Learn More

HSP70 Rabbit anti-Fish, Y. pseudotuberculosis, Polyclonal Antibody, Abnova™

Product Code. 16273330
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16273330 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16273330 Supplier Abnova Supplier No. PAB13575.200ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of HSP70.

Specifications

Antigen HSP70
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of HSP70.
Dilution Western Blot (1:15000) Immunoprecipitation (5 mg) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.4
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to C-terminus of salmonid Hsp70.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Fish
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with 200μL deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.