Learn More
CNN2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CNN2.
Brand: Abnova PAB26981.100uL
Description
The protein encoded by this gene, which can bind actin, calmodulin, troponin C, and tropomyosin, may function in the structural organization of actin filaments. The encoded protein could play a role in smooth muscle contraction and cell adhesion. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| CNN2 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| CNN2 | |
| CNN2 | |
| A synthetic peptide corresponding to human CNN2. | |
| 100 μL | |
| Primary | |
| CNN2 polyclonal antibody detects endogenous levels of CNN2 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of CNN2. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| Q99439 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1265 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only