Learn More
CBLC Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CBLC.
Brand: Abnova PAB1698.400uL
Description
CBL proteins, such as CBLC, are phosphorylated upon activation of a variety of receptors that signal via protein tyrosine kinases. Through interactions with proteins containing SRC (MIM 190090) homology-2 (SH2) and SH3 domains, CBL proteins modulate downstream cell signaling (Keane et al., 1999 [PubMed 10362357]).[supplied by OMIM
Specifications
| CBLC | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CBLC. | |
| In PBS (0.09% sodium azide) | |
| NP_036248;Q9ULV8 | |
| CBLC | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human CBLC. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| CBLC | |
| CBL-3/CBL-SL/RNF57 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 23624 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |