Learn More
CACNA1C Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CACNA1C.
Brand: Abnova PAB27743.50ug
Description
This gene encodes an alpha-1 subunit of a voltage-dependent calcium channel. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization. The alpha-1 subunit consists of 24 transmembrane segments and forms the pore through which ions pass into the cell. The calcium channel consists of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. There are multiple isoforms of each of these proteins, either encoded by different genes or the result of alternative splicing of transcripts. The protein encoded by this gene binds to and is inhibited by dihydropyridine. Alternative splicing results in many transcript variants encoding different proteins. [provided by RefSeq
Specifications
| CACNA1C | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CACNA1C. | |
| In PBS (0.09% sodium azide) | |
| Q13936 | |
| CACNA1C | |
| A synthetic peptide corresponding to 16 amino acid at internal region of human CACNA1C. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| CACNA1C | |
| CACH2/CACN2/CACNL1A1/CCHL1A1/CaV1.2/MGC120730/TS | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 775 | |
| Human | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only