missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB8A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55296-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to RAB8A.
The protein encoded by this gene is a member of the RAS superfamily which are small GTP/GDP-binding proteins with an average size of 200 amino acids. The RAS-related proteins of the RAB/YPT family may play a role in the transport of proteins from the endoplasmic reticulum to the Golgi and the plasma membrane. This protein shares 97%, 96%, and 51% similarity with the dog RAB8, mouse MEL, and mouse YPT1 proteins, respectively and contains the 4 GTP/GDP-binding sites that are present in all the RAS proteins. The putative effector-binding site of this protein is similar to that of the RAB/YPT proteins. However, this protein contains a C-terminal CAAX motif that is characteristic of many RAS superfamily members but which is not found in YPT1 and the majority of RAB proteins. Although this gene was isolated as a transforming gene from a melanoma cell line, no linkage between MEL and maligt melanoma has been demonstrable. This oncogene is located 800 kb distal to MY09B on chromosome 19p13.1.Specifications
| RAB8A | |
| Polyclonal | |
| Unconjugated | |
| RAB8A | |
| MEL, Oncogene c mel, RAB8, RAB8A, Ras related protein Rab 8A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117103, 17274, 4218 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35280, P55258, P61006 | |
| Rab8a | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction