missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB6A Recombinant Rabbit Monoclonal Antibody (HL1047)
GREENER_CHOICE

Product Code. 17718993
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17718993 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17718993 Supplier Invitrogen™ Supplier No. MA542374

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Rab proteins are also an integral part of endocytic pathways. Rab 6A is a ubiquitously expressed member of the Rab family of proteins and localizes to the Golgi membrane where it regulates retrograde transport from the late endosomes via the Golgi to endoplasmic reticulum (ER) pathway. Three isoforms exist due to alternative splicing events, namely the ubiquitously expressed isoforms Rab 6A' and Rab 6A, and the brain-specific isoform Rab 6B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB6A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1047
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene RAB6A
Gene Accession No. P20340, P35279, Q9WVB1
Gene Alias 2610028L11Rik; AA419671; MNCb-1660; Rab GTPase; Rab6; rab-6; RAB6, member RAS oncogene family; RAB6A; RAB6A, member RAS oncogene family; Rab6b; RAB6B, member RAS oncogene family; ras-related protein Rab-6A; RCO4-3
Gene Symbols RAB6A
Host Species Rabbit
Immunogen Full length human RAB6A recombinant protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19346, 5870, 84379
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.