missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB43 Antibody (5G4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00339122-M01
This item is not returnable.
View return policy
Description
RAB43 Monoclonal antibody specifically detects RAB43 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| RAB43 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| MGC90481, RAB11B, RAB41ISY1, RAB43, member RAS oncogene family, Ras-related protein Rab-41, ras-related protein Rab-43 | |
| RAB43 (NP_940892, 113 a.a. ∽ 210 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. IEDVRKYAGSNIVQLLIGNKSDLSELREVSLAEAQSLAEHYDILCAIETSAKDSSNVEEAFLRVATELIMRHGGPLFSEKSPDHIQLNSKDIGEGWGC | |
| 0.1 mg | |
| Signal Transduction | |
| 339122 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 5G4 | |
| Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| NP_940892 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction