Learn More
(R)-Usnic acid Sodium, TRC
Specifications
Specifications
| Chemical Name or Material | (R)-Usnic acid Sodium |
| CAS | 2796975-82-9 |
| Molecular Formula | C18 H15 Na O7 |
| Quantity | 500 mg |
| Synonym | sodium (R)-2,6-diacetyl-7,9-dihydroxy-8,9b-dimethyl-1-oxo-1,9b-dihydrodibenzo[b,d]furan-3-olate |
| InChI Formula | InChI=1S/C18H16O7.Na/c1-6-14(22)12(8(3)20)16-13(15(6)23)18(4)10(25-16)5-9(21)11(7(2)19)17(18)24;/h5,21-23H,1-4H3;/q;+1/p-1/t18-;/m0./s1 |
| SMILES | CC(=O)C1=C(O[Na])C=C2Oc3c(C(=O)C)c(O)c(C)c(O)c3[C@@]2(C)C1=O |
| IUPAC Name | [(9bR)-2,6-diacetyl-7,9-dihydroxy-8,9b-dimethyl-1-oxodibenzofuran-3-yl]oxysodium |
| Molecular Weight (g/mol) | 366.297 |
| Formula Weight | 366.072 |