Learn More
(R,S)-Anatabine, TRC
Product Code. 18013320
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18013320 100 mg 100mg
18042310 10 mg 10mg
18022920 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18013320 Supplier Toronto Research Chemicals Supplier No. TRCA637500100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2743-90-0
Molecular Formula C10 H12 N2
Molecular Weight (g/mol) 160.22
Synonym 2,3'-Bipyridine, 1,2,3,6-tetrahydro- (9CI, ACI), 1,2,3,6-Tetrahydro-2,3'-bipyridine (ACI), 2,3'-Bipyridine, 1,2,3,6-tetrahydro-, (±)- (ZCI), (RS)-Anatabine, (±)-1,2,3,6-Tetrahydro-2,3'-bipyridine, (±)-Anatabine, 1,2,3,6-Tetrahydro-2,3'-dipyridine, 3-(1,2,3,6-Tetrahydropyridin-2-yl)pyridine
IUPAC Name 3-(1,2,3,6-tetrahydropyridin-2-yl)pyridine
SMILES C1NC(CC=C1)c2cccnc2
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C10H12N2/c1-2-7-12-10(5-1)9-4-3-6-11-8-9/h1-4,6,8,10,12H,5,7H2
Formula Weight 160.1
Chemical Name or Material (+/-)-Anatabine