missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Rolipram, MedChemExpress

Product Code. 16477198
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16477198

Brand: Medchem Express HY16900A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (R)-(-)-Rolipram is the R-enantiomer of Rolipram. Rolipram is a selective inhibitor of phosphodiesterases PDE4 with IC50 of 3 nM, 130 nM and 240 nM for PDE4A, PDE4B, and PDE4D, respectively.

(R)-(-)-Rolipram is the R-enantiomer of Rolipram. Rolipram is a selective inhibitor of phosphodiesterases PDE4 with IC50 of 3 nM, 130 nM and 240 nM for PDE4A, PDE4B, and PDE4D, respectively.

Specifications

Chemical Name or Material (R)-(-)-Rolipram
CAS 85416-75-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H21NO3
Quantity 10 mg
Synonym (R)-Rolipram (-)-Rolipram
Solubility Information DMSO : 100 mg/mL (363.19 mM; Need ultrasonic)
SMILES O=C1NC[C@@H](C2=CC=C(OC)C(OC3CCCC3)=C2)C1
Molecular Weight (g/mol) 275.34
Formula Weight 275.34
Percent Purity 98.6%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.