Learn More
(R,R)-BD-AcAc 2, MedChemExpress
Product Code. 16416746
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 16416746
Brand: Medchem Express HY15344
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress (R,R)-BD-AcAc 2 ((R,R)-Ketone Ester), a ketone monoester, can be used as a source of oral nutritional ketones.
(R,R)-BD-AcAc 2 can elevate plasma levels of β-hydroxybutyrate and acetoacetate, blood glucose, blood Na+ levels and blood creatinine levels after oral administration in mice. (R,R)-BD-AcAc 2 can partly prevent muscle weakness in septic mice. (R,R)-BD-AcAc 2 has potential to improve exercise performance and endurance in animal body. (R,R)-BD-AcAc 2 can also be used to research Parkinson’s disease or diabetes.
Specifications
Chemical Name or Material (R,R)-BD-AcAc 2
Physical Form Viscous Liquid
CAS 1208313-97-6
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H16O4
For Use With (Application) Neuroscience-Neurodegeneration
Synonym (R,R)-Ketone Ester
Solubility Information H2O : 100 mg/mL (567.50 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (567.50 mM; Need ultrasonic)
SMILES C[C@@H](O)CC(OCC[C@H](O)C)=O
Molecular Weight (g/mol) 176.21
Formula Weight 176.21
Percent Purity 95.0%
Grade Research