Learn More
(R)-N-(1-(Naphthalen-1-yl)ethyl)-3-(3-(trifluoromethyl)phenyl)propanamide, TRC
Product Code. 30479035
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479035 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30479035 Supplier Toronto Research Chemicals Supplier No. TRCN37848525MG
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (R)-N-(1-(Naphthalen-1-yl)ethyl)-3-(3-(trifluoromethyl)phenyl)propanamide
CAS 1005450-55-4
Molecular Formula C22 H20 F3 N O
Quantity 25 mg
Synonym Benzenepropanamide','N-[(1R)-1-(1-naphthalenyl)ethyl]-3-(trifluoromethyl)-, N-[(1R)-1-(1-Naphthalenyl)ethyl]-3-(trifluoromethyl)benzenepropanamide, N-[1-(R)-(1-Naphthyl)ethyl]-3-[3-trifluoromethylphenyl]propanamide
InChI Formula InChI=1S/C22H20F3NO/c1-15(19-11-5-8-17-7-2-3-10-20(17)19)26-21(27)13-12-16-6-4-9-18(14-16)22(23,24)25/h2-11,14-15H,12-13H2,1H3,(H,26,27)/t15-/m1/s1
SMILES C[C@@H](NC(=O)CCc1cccc(c1)C(F)(F)F)c2cccc3ccccc23
IUPAC Name N-[(1R)-1-naphthalen-1-ylethyl]-3-[3-(trifluoromethyl)phenyl]propanamide
Molecular Weight (g/mol) 371.4
Formula Weight 371.1497