Learn More
R&D Systems™ Mouse Serpin F1/PEDF Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 8295-SF-025
Description
The Recombinant Mouse Serpin F1/PEDF Protein is derived from NS0. The Recombinant Mouse Serpin F1/PEDF Protein has been validated for the following applications: Bioactivity.Specifications
| BAA31978.1 | |
| 20317 | |
| Serpin F1/PEDF | |
| SDS-PAGE: 50-56kDa, reducing conditions | |
| Mouse myeloma cell line,NS0-derived mouse Serpin F1/PEDF protein Gln20-Thr417,with a C-terminal 6-His tag | |
| RUO | |
| Carrier Protein | |
| Yes |
| Lyophilized from a 0.2μm filtered solution in PBS and NaCl | |
| 45kDa | |
| Reconstitute at 250μg/mL in sterile PBS. | |
| 25μg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| No results obtained: Gln20 inferred from enzymatic pyroglutamate treatment revealing Asn21 | |
| Unconjugated |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.