Learn More
R-Mirtazapine-d3, TRC
Specifications
Specifications
| Chemical Name or Material | R-Mirtazapine-d3 |
| CAS | 2751591-56-5 |
| Molecular Formula | C17H16D3N3 |
| Quantity | 25 mg |
| Synonym | (14bR)-1,2,3,4,10,14b-Hexahydro-2-methyl-pyrazino[2,1-a]pyrido[2,3-c][2]benzazepine, (R)-1,2,3,4,10,14b-Hexahydro-2-methyl-pyrazino[2,1-a]pyrido[2,3-c][2]benzazepine-d3, (-)-Mirtazapine-d3, (R)-6-Azamianserin-d3, (R)-Mirtazapine-d3, (R)-Org 3770-d3, Org 44-19-d3, R-(-)-Mirtazapine-d3 |
| InChI Formula | InChI=1S/C17H19N3/c1-19-9-10-20-16(12-19)15-7-3-2-5-13(15)11-14-6-4-8-18-17(14)20/h2-8,16H,9-12H2,1H3/t16-/m0/s1/i1D3 |
| SMILES | [2H]C([2H])([2H])N(C1)CCN2[C@@H]1C(C=CC=C3)=C3CC4=C2N=CC=C4 |
| IUPAC Name | (R)-2-(methyl-d3)-1,2,3,4,10,14b-hexahydrobenzo[c]pyrazino[1,2-a]pyrido[3,2-f]azepine |
| Molecular Weight (g/mol) | 268.37 |
| Formula Weight | 268.18 |