Learn More
R&D Systems™ Human GABA-A R delta (NP_000806) VersaClone cDNA
Product Code. 18793379
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18793379 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18793379 Supplier R&D Systems™ Supplier No. RDC1047
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
gamma-Aminobutyric Acid Type A Receptor, delta Polypeptide
The Human GABA-A R delta (NP_000806) VersaClone cDNA clone contains the complete ORF for GABA-A R delta, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The GABA-A R delta insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans