missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse 4-1BB Ligand/TNFSF9 Protein

Product Code. 18374947
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18374947

Brand: R&D Systems™ 12464L025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity

Measured by its ability to co-stimulate IL-2 secretion by mouse T cells in the presence of anti-CD3. The ED50 for this effect is 0.008-0.05 μg/mL in the presence of a Mouse Anti-polyHistidine Monoclonal Antibody (Catalog # MAB050 ). Measured by its ability to bind Recombinant Mouse 4-1BB/TNFRSF9/CD137 Fc Chimera (Catalog # 937-4B ) in a functional ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P41274
For Use With (Application) Bioactivity, Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 8744
Molecular Weight (g/mol) M.W. (Observed): 41-44 kDa, reducing conditions; M.W. (theoretical): 25.7 kDa
Quantity 25 μg
Source Mouse myeloma cell line, NS0-derived mouse 4-1 BB Ligand/TNFSF9 protein Met 10-His tag GGGSGGGSGGGSIEGR Mouse 4-1 BB Ligand (Arg104-Glu309) (N-terminus) Accession # P41274 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol Tnfsf9
Product Type 4-1BB Ligand/TNFSF9
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein 4-1 BB Ligand/TNFSF9
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.