Learn More
R&D Systems™ Human IL-23R (AAM44229) VersaClone cDNA
IL-23R; interleukin 23 receptor; interleukin-23 receptor
Brand: R&D Systems™ RDC1255
Description
The Human IL-23 R (AAM44229) VersaClone cDNA clone contains the complete ORF for IL-23R, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The IL-23R insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature
Specifications
| hIL23R | |
| 10 μg at 0.2 μg/μL | |
| 1930bp | |
| IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor | |
| Human, Mouse |
| AAM44229 | |
| cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
| 149233, 209590 | |
| Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
| 10 μg |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Reasearch Use Only; Not for Use in Humans.