Learn More
R-(+)-Cotinine, TRC
Specifications
Specifications
| Chemical Name or Material | R-(+)-Cotinine |
| CAS | 32162-64-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 H12 N2 O |
| Quantity | 10 mg |
| Synonym | 2-Pyrrolidinone, 1-methyl-5-(3-pyridinyl)-, (5R)-, 2-Pyrrolidinone, 1-methyl-5-(3-pyridinyl)-, (R)- (9CI), Cotinine, (+)- (8CI), (5R)-1-Methyl-5-(3-pyridinyl)-2-pyrrolidinone, (+)-Cotinine, (R)-Cotinine, Ba 2701, R-(+)-Cotinine, (5R)-1-Methyl-5-(pyridin-3-yl)pyrrolidin-2-one |
| InChI Formula | InChI=1S/C10H12N2O/c1-12-9(4-5-10(12)13)8-3-2-6-11-7-8/h2-3,6-7,9H,4-5H2,1H3/t9-/m1/s1 |
| SMILES | CN1[C@H](CCC1=O)c2cccnc2 |
| IUPAC Name | (5R)-1-methyl-5-pyridin-3-ylpyrrolidin-2-one |
| Molecular Weight (g/mol) | 176.22 |
| Show More |