Learn More
(R)-Bupivacaine, TRC
Product Code. 30341590
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30341590 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30341590 Supplier Toronto Research Chemicals Supplier No. TRCB45122510MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (R)-Bupivacaine
CAS 27262-45-9
Molecular Formula C18H28N2O
Quantity 10 mg
Synonym 2-Piperidinecarboxamide, 1-butyl-N-(2,6-dimethylphenyl)-, (2R)-, 2-Piperidinecarboxamide, 1-butyl-N-(2,6-dimethylphenyl)-, (R)-, 2',6'-Pipecoloxylidide, 1-butyl-, (+)- (8CI), (2R)-1-Butyl-N-(2,6-dimethylphenyl)-2-piperidinecarboxamide, (+)-(R)-Bupivacaine, (+)-Bupivacaine, (R)-(+)-Bupivacaine, (R)-Bupivacaine, D-(+)-Bupivacaine, d-Bupivacaine
InChI Formula InChI=1S/C18H28N2O/c1-4-5-12-20-13-7-6-11-16(20)18(21)19-17-14(2)9-8-10-15(17)3/h8-10,16H,4-7,11-13H2,1-3H3,(H,19,21)/t16-/m1/s1
SMILES CCCCN1CCCC[C@@H]1C(=O)NC2=C(C=CC=C2C)C
IUPAC Name (R)-1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide
Molecular Weight (g/mol) 288.4
Formula Weight 288.2202