Learn More
(R)-(-)-Bromo Dragonfly Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (R)-(-)-Bromodragonfly HCl |
| CAS | 332012-24-5 |
| Molecular Formula | C13H13BrClNO2 |
| Quantity | 10 mg |
| Synonym | Benzo[1,2-b:4,5-b']difuran-4-ethanamine, 8-bromo-α-methyl-, hydrochloride, (αR)- (9CI), (R)-1-(4-Bromofuro[2,3-f][1]benzofuran-8-yl)propan-2-amine Hydrochloride, (R)-(-)-Bromodragonfly HCl |
| InChI Formula | InChI=1S/C13H12BrNO2.ClH/c1-7(15)6-10-8-2-4-17-13(8)11(14)9-3-5-16-12(9)10;/h2-5,7H,6,15H2,1H3;1H/t7-;/m1./s1 |
| SMILES | Cl.C[C@@H](N)Cc1c2occc2c(Br)c3occc13 |
| IUPAC Name | (2R)-1-(4-bromofuro[2,3-f][1]benzofuran-8-yl)propan-2-amine;hydrochloride |
| Molecular Weight (g/mol) | 330.6 |