Learn More
(R)-Baclofen, MedChemExpress
Product Code. 16483307
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16483307
Brand: Medchem Express HY17354
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress (R)-Baclofen (Arbaclofen) is a selective GABAB receptor agonist.
(R)-Baclofen (Arbaclofen) is a selective GABAB receptor agonist.
Specifications
Chemical Name or Material (R)-Baclofen
Physical Form Powder
CAS 69308-37-8
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C10H12ClNO2
For Use With (Application) Neuroscience-Neuromodulation
Synonym Arbaclofen STX209
Solubility Information H2O : 5 mg/mL (23.40 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (4.68 mM; ultrasonic and warming and heat to 80°C)
SMILES ClC1=CC=C([C@](CC(O)=O)([H])CN)C=C1
Molecular Weight (g/mol) 213.66
Formula Weight 213.66
Percent Purity 98.72%
Grade Research